Anti-Caspase 9, active Rabbit Polyclonal Antibody
Lieferant: US Biological
C2088-13
USBIC2088-13EA 741 EUR
USBIC2088-13
Anti-Caspase 9, active Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 10-20 ?g/ml; Immunoprecipitation: 10-20 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Caspase 9, active
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat