Anti-Caspase-9, active Rabbit Polyclonal Antibody

Lieferant: US Biological
C2088-13A
USBIC2088-13AEA 539 EUR
USBIC2088-13A
Anti-Caspase-9, active Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: Caspase-9, active
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR