Anti-Caspase-9, Active Rabbit Polyclonal Antibody

Lieferant: US Biological
C2088-51D
USBIC2088-51DEA 572 EUR
USBIC2088-51D
Anti-Caspase-9, Active Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml; Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: Caspase-9, Active
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR