Anti-CTSC Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
C2097-16B
USBIC2097-16BEA 626 EUR
USBIC2097-16B
Anti-CTSC Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CTSC (cathepsin C)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR