Anti-CD3, epsilon Rat Monoclonal Antibody [clone: 5K4]

Lieferant: US Biological
C2254-11LX
USBIC2254-11LXEA 425 EUR
USBIC2254-11LX
Anti-CD3, epsilon Rat Monoclonal Antibody [clone: 5K4]
Antikörper
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2c Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR