Anti-CD3 Mouse Monoclonal Antibody [clone: 5K205]

Lieferant: US Biological
C2254-56JX-100
USBIC2254-56JX-100EA 250 EUR
USBIC2254-56JX-100
Anti-CD3 Mouse Monoclonal Antibody [clone: 5K205]
Antikörper
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 5K205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR