Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Texas Red®) [clone: 53-6.7]

Lieferant: US Biological
C2259-28E
USBIC2259-28EEA 487 EUR
USBIC2259-28E
Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Texas Red®) [clone: 53-6.7]
Antikörper
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR