Anti-CD9 Mouse Monoclonal Antibody [clone: CLB-47]

Lieferant: US Biological
C2260-06L
USBIC2260-06LEA 704 EUR
USBIC2260-06L
Anti-CD9 Mouse Monoclonal Antibody [clone: CLB-47]
Antikörper
Type: Primary
Antigen: CD9 (CD9 molecule)
Clonality: Monoclonal
Clone: CLB-47
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR