Anti-ITGAM Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B618]

Lieferant: US Biological
C2262-35C
USBIC2262-35CEA 701 EUR
USBIC2262-35C
Anti-ITGAM Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B618]
Antikörper
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: Monoclonal
Clone: 10B618
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR