Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 5K8]

Lieferant: US Biological
C2262-35Q
USBIC2262-35QEA 419 EUR
USBIC2262-35Q
Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 5K8]
Antikörper
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: Monoclonal
Clone: 5K8
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR