Anti-ITGAM Rat Monoclonal Antibody [clone: 3H977]

Lieferant: US Biological
C2262-37A
USBIC2262-37AEA 918 EUR
USBIC2262-37A
Anti-ITGAM Rat Monoclonal Antibody [clone: 3H977]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: Monoclonal
Clone: 3H977
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR