Anti-ITGAM Mouse Monoclonal Antibody [clone: 6A244]

Lieferant: US Biological
C2262-37J
USBIC2262-37JEA 937 EUR
USBIC2262-37J
Anti-ITGAM Mouse Monoclonal Antibody [clone: 6A244]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: Monoclonal
Clone: 6A244
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR