Anti-CD11b Rat Monoclonal Antibody [clone: M1/70]

Lieferant: US Biological
C2262-43K-50
USBIC2262-43K-50EA 311 EUR
USBIC2262-43K-50
Anti-CD11b Rat Monoclonal Antibody [clone: M1/70]
Antikörper
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Human, Monkey, Mouse, Rabbit,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR