Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5®) [clone: MB19-1]

Lieferant: US Biological
C2269-73F
USBIC2269-73FEA 446 EUR
USBIC2269-73F
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5®) [clone: MB19-1]
Antikörper
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Monoclonal
Clone: MB19-1
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR