Anti-FCER2 Mouse monoclonal antibody [clone: 5K232]
Lieferant: US Biological
C2273-89W
USBIC2273-89WEA 320 EUR
USBIC2273-89W
Anti-FCER2 Mouse monoclonal antibody [clone: 5K232]
Antikörper
Type: Primary
Antigen: FCER2 (Fc fragment of IgE, low affinity II, receptor for (CD23))
Clonality: Monoclonal
Clone: 5K232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human