Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 3C7]

Lieferant: US Biological
C2275-82F
USBIC2275-82FEA 419 EUR
USBIC2275-82F
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 3C7]
Antikörper
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: Monoclonal
Clone: 3C7
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR