Anti-DPP4 Mouse Monoclonal Antibody [clone: 202.36]

Lieferant: US Biological
C2277-07J
USBIC2277-07JEA 506 EUR
USBIC2277-07J
Anti-DPP4 Mouse Monoclonal Antibody [clone: 202.36]
Antikörper
Type: Primary
Antigen: DPP4 (dipeptidyl-peptidase 4)
Clonality: Monoclonal
Clone: 202.36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR