Anti-CD28 Mouse Monoclonal Antibody [clone: 204.12]

Lieferant: US Biological
C2380-01K
USBIC2380-01KEA 843 EUR
USBIC2380-01K
Anti-CD28 Mouse Monoclonal Antibody [clone: 204.12]
Antikörper
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: Monoclonal
Clone: 204.12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR