Anti-CD28 Hamster Monoclonal Antibody [clone: 5K12]

Lieferant: US Biological
C2380-08GX
USBIC2380-08GXEA 460 EUR
USBIC2380-08GX
Anti-CD28 Hamster Monoclonal Antibody [clone: 5K12]
Antikörper
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: Monoclonal
Clone: 5K12
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR