Anti-TNFRSF8 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K13]

Lieferant: US Biological
C2382-14C
USBIC2382-14CEA 473 EUR
USBIC2382-14C
Anti-TNFRSF8 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K13]
Antikörper
Type: Primary
Antigen: TNFRSF8 (tumor necrosis factor receptor superfamily, member 8)
Clonality: Monoclonal
Clone: 5K13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR