Anti-CD38 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5K14]
Lieferant: US Biological
C2390-13E
USBIC2390-13EEA 492 EUR
USBIC2390-13E
Anti-CD38 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5K14]
Antikörper
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: Monoclonal
Clone: 5K14
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse