Anti-CD38 Rat Monoclonal Antibody [clone: 5K253]

Lieferant: US Biological
C2390-25AX
USBIC2390-25AXEA 540 EUR
USBIC2390-25AX
Anti-CD38 Rat Monoclonal Antibody [clone: 5K253]
Antikörper
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: Monoclonal
Clone: 5K253
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR