Anti-CD44 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H174]

Lieferant: US Biological
C2398-06H
USBIC2398-06HEA 708 EUR
USBIC2398-06H
Anti-CD44 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H174]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 7H174
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR