Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]

Lieferant: US Biological
C2399-02K
USBIC2399-02KEA 809 EUR
USBIC2399-02K
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]
Antikörper
Do not freeze!

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone: C11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR