Anti-CD46 Mouse Monoclonal Antibody [clone: MEM-258]

Lieferant: US Biological
C2401-02K
USBIC2401-02KEA 633 EUR
USBIC2401-02K
Anti-CD46 Mouse Monoclonal Antibody [clone: MEM-258]
Antikörper
Type: Primary
Antigen: CD46 (CD46 molecule, complement regulatory protein)
Clonality: Monoclonal
Clone: MEM-258
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR