Anti-CD46 Mouse Monoclonal Antibody [clone: 169-1-E4.3]

Lieferant: US Biological
C2401-02L
USBIC2401-02LEA 439 EUR
USBIC2401-02L
Anti-CD46 Mouse Monoclonal Antibody [clone: 169-1-E4.3]
Antikörper
Type: Primary
Antigen: CD46 (CD46 molecule, complement regulatory protein)
Clonality: Monoclonal
Clone: 169-1-E4.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR