Anti-CD49d Rat Monoclonal Antibody [clone: 2B2.32]

Lieferant: US Biological
C2404-42A-100
USBIC2404-42A-100EA 289 EUR
USBIC2404-42A-100
Anti-CD49d Rat Monoclonal Antibody [clone: 2B2.32]
Antikörper
Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: 2B2.32
Conjugation: Unconjugated
Epitope:
Host: IgG2b kappa
Isotype: Rat
Reactivity: Human, Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR