Anti-ITGA4 Rat Monoclonal Antibody [clone: 2B2.32]
Lieferant: US Biological
C2404-42A-500
USBIC2404-42A-500EA 552 EUR
USBIC2404-42A-500
Anti-ITGA4 Rat Monoclonal Antibody [clone: 2B2.32]
Antikörper
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: Monoclonal
Clone: 2B2.32
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Human, Mouse