Anti-NCAM1 Mouse Monoclonal Antibody [clone: 5F47]

Lieferant: US Biological
C2410-59
USBIC2410-59EA 338 EUR
USBIC2410-59
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 5F47]
Antikörper
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 5F47
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR