Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5®) [clone: 5K269]

Lieferant: US Biological
C2410-76F
USBIC2410-76FEA 492 EUR
USBIC2410-76F
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5®) [clone: 5K269]
Antikörper
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 5K269
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR