Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5F69]

Lieferant: US Biological
C2412-07C
USBIC2412-07CEA 378 EUR
USBIC2412-07C
Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5F69]
Antikörper
Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: Monoclonal
Clone: 5F69
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR