Anti-FCGR1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H102]

Lieferant: US Biological
C2419-06K
USBIC2419-06KEA 539 EUR
USBIC2419-06K
Anti-FCGR1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H102]
Antikörper
Do not freeze!

Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: Monoclonal
Clone: 7H102
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR