Anti-CEACAM8 Mouse monoclonal antibody [clone: 1.BB.530]
Lieferant: US Biological
C2421-01C
USBIC2421-01CEA 796 EUR
USBIC2421-01C
Anti-CEACAM8 Mouse monoclonal antibody [clone: 1.BB.530]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100; Immunochemistry: 1:25-1-100
Type: Primary
Antigen: CEACAM8 (carcinoembryonic antigen-related cell adhesion molecule 8)
Clonality: Monoclonal
Clone: 1.BB.530
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human