Anti-ENG Mouse monoclonal antibody [clone: MEM-226]
Lieferant: US Biological
C2446-51E
USBIC2446-51EEA 634 EUR
USBIC2446-51E
Anti-ENG Mouse monoclonal antibody [clone: MEM-226]
Antikörper
Type: Primary
Antigen: ENG (endoglin)
Clonality: Monoclonal
Clone: MEM-226
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgGa
Reactivity: Human