Anti-ENG Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-226]

Lieferant: US Biological
C2446-51F
USBIC2446-51FEA 742 EUR
USBIC2446-51F
Anti-ENG Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-226]
Antikörper
Recommended dilution: Flow Cytometry: 20?l reagent/100?l sample

Type: Primary
Antigen: ENG (endoglin)
Clonality: Monoclonal
Clone: MEM-226
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR