Anti-CD152, Fab Monomer Mouse Monoclonal Antibody [clone: 5D3]

Lieferant: US Biological
C2548-83G5
USBIC2548-83G5EA 1350 EUR
USBIC2548-83G5
Anti-CD152, Fab Monomer Mouse Monoclonal Antibody [clone: 5D3]
Antikörper
Type: Primary
Antigen: CD152, Fab Monomer
Clonality: Monoclonal
Clone: 5D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR