Anti-CDC25A Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
C2560-01G-BIOT
USBIC2560-01G-BIOTEA 884 EUR
USBIC2560-01G-BIOT
Anti-CDC25A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR