Anti-CDK1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
C2571-07B-BIOT
USBIC2571-07B-BIOTEA 884 EUR
USBIC2571-07B-BIOT
Anti-CDK1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human