Anti-Ceramide Mouse monoclonal antibody [clone: 6D311]
Lieferant: US Biological
C2777-95
USBIC2777-95EA 910 EUR
USBIC2777-95
Anti-Ceramide Mouse monoclonal antibody [clone: 6D311]
Antikörper
Recommended dilution: ELISA: 1:10; Flow Cytometry: 20?l/test; Immunochemistry: 1:10
Type: Primary
Antigen: Ceramide
Clonality: Monoclonal
Clone: 6D311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Dog, Goat, Monkey, Pig, Rat, Sheep