Anti-CHD2 Rabbit Polyclonal Antibody
Lieferant: US Biological
C3875-25
USBIC3875-25EA 632 EUR
USBIC3875-25
Anti-CHD2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:10-1:1000; Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: CHD2 (chromodomain helicase DNA binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human