Anti-Chemokine Receptor FKSG80 Rabbit Polyclonal Antibody

Lieferant: US Biological
C3915-02A
USBIC3915-02AEA 648 EUR
USBIC3915-02A
Anti-Chemokine Receptor FKSG80 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:60; Immunochemistry: 1:60

Type: Primary
Antigen: Chemokine Receptor FKSG80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR