Anti-Chlamydia sp. LPS Mouse Monoclonal Antibody [clone: 10B2222]

Lieferant: US Biological
C4250-42B
USBIC4250-42BEA 679 EUR
USBIC4250-42B
Anti-Chlamydia sp. LPS Mouse Monoclonal Antibody [clone: 10B2222]
Antikörper
Recommended dilution: ELISA: 1:2000

Type: Primary
Antigen: Chlamydia sp. LPS
Clonality: Monoclonal
Clone: 10B2222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR