Anti-CHN2 Goat Polyclonal Antibody

Lieferant: US Biological
C5038-20
USBIC5038-20EA 601 EUR
USBIC5038-20
Anti-CHN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000

Type: Primary
Antigen: CHN2 (chimerin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR