Anti-PTK7 Rabbit Polyclonal Antibody

Lieferant: US Biological
C5042-09A
USBIC5042-09AEA 667 EUR
USBIC5042-09A
Anti-PTK7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PTK7 (protein tyrosine kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR