Anti-PTK7 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
C5042-09A-BIOT
USBIC5042-09A-BIOTEA 884 EUR
USBIC5042-09A-BIOT
Anti-PTK7 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PTK7 (protein tyrosine kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR