Anti-Cholesterol Acyltransferase 1 Rabbit Polyclonal Antibody
Lieferant: US Biological
C5045-33C
USBIC5045-33CEA 614 EUR
USBIC5045-33C
Anti-Cholesterol Acyltransferase 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:400
Type: Primary
Antigen: Cholesterol Acyltransferase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Mouse