Anti-Cholesterol Acyltransferase 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
C5045-35
USBIC5045-35EA 652 EUR
USBIC5045-35
Anti-Cholesterol Acyltransferase 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:1000;

Type: Primary
Antigen: Cholesterol Acyltransferase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR