Anti-Cholesterol Esterase Rabbit Polyclonal Antibody

Lieferant: US Biological
C5047-05A
USBIC5047-05AEA 809 EUR
USBIC5047-05A
Anti-Cholesterol Esterase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cholesterol Esterase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR