Anti-Cholinergic Receptor, Nicotinic, alpha 5 Rabbit Polyclonal Antibody

Lieferant: US Biological
C5062-74B
USBIC5062-74BEA 602 EUR
USBIC5062-74B
Anti-Cholinergic Receptor, Nicotinic, alpha 5 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: Cholinergic Receptor, Nicotinic, alpha 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR