Anti-cholinergic receptor, nicotinic, beta 2 Rabbit Polyclonal Antibody

Lieferant: US Biological
C5062-74X
USBIC5062-74XEA 829 EUR
USBIC5062-74X
Anti-cholinergic receptor, nicotinic, beta 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:20000

Type: Primary
Antigen: cholinergic receptor, nicotinic, beta 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR